Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG F(ab) Antibody (RMG05), Biotin, R&D Systems™
SDP

Catalog No. NBP262016B Shop All Novus Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP262016B 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP262016B Supplier R&D Systems Supplier No. NBP262016B
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Goat Monoclonal Antibody

IgG F(ab) Monoclonal specifically detects IgG F(ab) in Mouse samples. It is validated for ELISA.

Specifications

Antigen IgG F(ab)
Applications ELISA
Classification Monoclonal
Clone RMG05
Concentration 1 mg/mL
Conjugate Biotin
Dilution Flow Cytometry, ELISA 0.05ug/ml - 1ug/ml, Immunohistochemistry 0.5 - 2 ug/mL, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunocytochemistry 0.5 - 2 ug/mL
Formulation PBS, 50% Glycerol, 1% BSA with 0.09% Sodium Azide
Host Species Goat
Immunogen Mouse IgG
Purification Method Protein G purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Secondary
Test Specificity This antibody reacts to the Fab region of Mouse IgG, including IgG1, IgG2a, IgG2b, and IgG3. No cross reactivity with IgM, IgA, IgE, human IgG, rat IgG, or rabbit IgG.
Target Species Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.