Learn More
QA Bio Inc Ludger IgG N-Glycan Library, Procainamide labeled
Supplier:  QA Bio Inc CPROCIGG02
Encompass
A mixture of fucosylated, bi-antennary glycan standards with variable sialylation released from human IgG antibody glycoprotein. • Sources: The IgG glycan library in these products are released from an IgG standard that is purified from human serum. IgG exists in a variety of glycoforms containing bi-antennary oligosaccharides with variable sialylation. • Form: Dry. Dried by centrifugal evaporation from an aqueous solution. • Amount supplied: CLIBN-IGG-01 25 µgs – unlabeled • CAB-IGG-01 200 pmol – 2-AB labeled • CAPTS-IGG-01 10 pmol – APTS labeled • CPROC-IGG-02 50 pmol – procainamide labeled • CPM-IGG-01 20 MS runs – permethylated • CPM-C13-IGG-01 20 MS runs – permethylated with heavy (13C) MeI • Purity: > 90% pure as assessed by a combination of 1 H-NMR and HPLC, and CE
Catalog No. 50-223-6613
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.