Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgG Goat anti-Rabbit, AF 488, Polyclonal, Southern Biotech™

Goat Polyclonal Secondary Antibody
Supplier: Southern Biotechnology Associates 403030
Description
SouthernBiotech is a leading provider of polyclonal secondary antibodies offering high sensitivity and low background for use in a variety of applications.
Specifications
IgG | |
Polyclonal | |
Alexa Fluor 488 | |
Goat | |
Affinity Purified | |
RUO | |
Rabbit | |
IgG |
Flow Cytometry, ELISA, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
1.0mg/mL | |
Supplied in PBS/NaN3 | |
Rabbit IgG | |
1.0 mg | |
Secondary | |
Store at 2-8°C; Protect from light |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction