Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgG Goat F(ab')2 anti-Rat, HRP, Polyclonal, Southern Biotech™

Goat F(ab')2 Polyclonal Secondary Antibody
Supplier: Southern Biotechnology Associates 303205
Description
SouthernBiotech is a leading provider of polyclonal secondary antibodies offering high sensitivity and low background for use in a variety of applications.
Specifications
IgG | |
Polyclonal | |
HRP | |
Goat | |
Affinity Purified | |
RUO | |
Rat | |
IgG |
ELISA | |
Lot specific | |
Supplied in a stock solution in 50% glycerol/50% PBS, pH 7.4; No preservative added | |
Rat IgG | |
1.0 mL | |
Secondary | |
Store at 2-8°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction