Learn More
IgG Goat, LE/AF (Low Endotoxin/Azide Free), Polyclonal, Isotype Control, Southern Biotech™
Shop All Southern Biotechnology Associates Products

Description
SouthernBiotech isotype controls and purified immunoglobulins are useful as negative controls and standards in a variety of immunoassays.
Specifications
Specifications
| Applications | Blocking Assay, ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), In vitro Assay |
| Classification | Low Endotoxin/Azide-free (LE/AF) |
| Concentration | 0.5mg/mL |
| Formulation | Supplied in PBS |
| Host Species | Goat |
| Quantity | 0.5 mg |
| Regulatory Status | RUO |
| Primary or Secondary | Primary |
| Content And Storage | Store at -20°C |
| Isotype | IgG |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.