Learn More
IgG Goat F(ab')2, R-PE, Polyclonal, Isotype Control, Southern Biotech™
Goat F(ab')2 Polyclonal Isotype Control
Supplier: Southern Biotechnology Associates 011009
Description
SouthernBiotech isotype controls and purified immunoglobulins are useful as negative controls and standards in a variety of immunoassays.
Specifications
| Flow Cytometry, ELISA | |
| Lot specific | |
| Supplied in PBS/NaN3 and a stabilizing agent | |
| 100 Tests | |
| Primary | |
| IgG |
| Polyclonal | |
| R-PE | |
| Goat | |
| RUO | |
| Store at 2-8°C; Protect from light; Do not freeze |