Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG Goat, TRITC (Rhodamine), Polyclonal, Isotype Control, Southern Biotech™
SDP

Catalog No. OB010903 Shop All Southern Biotechnology Associates Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OB010903 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OB010903 Supplier Southern Biotechnology Associates Supplier No. 010903
Add to Cart
Add to Cart

Goat Polyclonal Isotype Control

SouthernBiotech isotype controls and purified immunoglobulins are useful as negative controls and standards in a variety of immunoassays.

Specifications

Applications ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration Lot specific
Conjugate TRITC
Formulation Supplied in PBS/NaN3
Host Species Goat
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage Store at 2-8°C; Protect from light
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.