Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG (H+L) Rabbit anti-Mouse, R-Phycoerythrin, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB002845 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1000 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB002845 1000 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB002845 Supplier Bio-techne Sales Corp Supplier No. NBP2685011000UL

Rabbit Polyclonal Antibody

IgG (H+L) Polyclonal specifically detects IgG (H+L) in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen IgG (H+L)
Applications Flow Cytometry
Classification Polyclonal
Concentration LYOPH
Conjugate R-PE
Dilution Flow Cytometry 1:20 - 1:100
Formulation PBS, 1% BSA, 5% Sucrose
Host Species Rabbit
Immunogen Mouse IgG
Quantity 1000 μL
Reconstitution Reconstitute with 1.0 mL distilled water. Store at 4°C.
Target Species Mouse
Content And Storage Store at 4°C. Do not freeze.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.