Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ IgG (Human) ELISA Kit
Click to view available options
Quantity:
1 Kit
96 Assays
Sample Type:
Cell Culture, Milk, Plasma, Saliva, Serum, Urine
Plasma, Serum
Sample Volume:
100 μL
50 μL
Description
- Sample Type: Plasma, Serum
- Sample Volume: 100μL
Quantification
Specifications
Specifications
Conjugate | Biotin |
Cross Reactivity | Human |
Description | IgG (Human) ELISA Kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human IgG. |
Detection Limit | 150pg/mL |
Detection Method | Colorimetric |
Includes | 96-well Antibody coated microwell, 25mL Wash Buffer 20X, 2 vials standard, 30mL assay diluent A, 15mL assay diluent B, 2 vials detection Antibody MSP, 8μL HRP Streptavidian concentrate, 12mL TMB one step Substrate reagent, 8mL stop solut |
Quantity | 96 Assays |
Sample Type | Plasma, Serum |
Sample Volume | 100 μL |
Content And Storage | Store the kit at -20°C. For best results, please store the components as described in the protocol. Avoid repeated freezing and thawing. |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction