Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat anti-Human IgG/IgM (H+L) Secondary Antibody, HRP (Horseradish Peroxidase), Novus Biologicals™
SDP

Goat Polyclonal Antibody

Supplier:  Novus Biologicals NBP266687

Catalog No. NBP266687


Only null left
Add to Cart

Description

Description

IgG/IgM (H+L) Polyclonal specifically detects IgG/IgM (H+L) in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry.
Specifications

Specifications

IgG/IgM(H+L)
Polyclonal
HRP
PBS, 0.2% BSA with 0.05% Pro-Clean 400
Human IgG+IgM-heavy and light chain
1 mL
Human
IgG
ELISA, Immunocytochemistry, Immunohistochemistry, Western Blot
1mg/mL
Western Blot 1:5000 - 1:50000, ELISA 1:10000 - 1:00000, Immunohistochemistry 1:200 - 1:5000, Immunocytochemistry 1:200 - 1:5000
Goat
Affinity Purified
Secondary
Store at 4°C. Do not freeze.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.