Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG Rabbit, Alexa Fluor 750, Clone: 60024B, Isotype Control, R&D Systems™

Catalog No. IC1051S
Change view
Click to view available options
Quantity:
200 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC1051S 200 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC1051S Supplier R&D Systems Supplier No. IC1051S
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

IgG Monoclonal specifically detects IgG in Rabbit samples. It is validated for Control.

Specifications

Antigen IgG
Classification Monoclonal
Clone 60024B
Conjugate Alexa Fluor 750
Dilution Control 5 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide.
Gene Alias Immunoglobulin G, ImmunoglobulinG
Host Species Rabbit
Immunogen KLH
Purification Method Protein A or G purified from cell culture supernatant
Quantity 200 Tests
Regulatory Status RUO
Gene ID (Entrez) 3500
Test Specificity Selected for use as rabbit IgG isotype control for flow cytometry applications.
Target Species Rabbit
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.