Learn More
IgG Rabbit, APC, Clone: 60024B, Isotype Control, R&D Systems™
Rabbit Monoclonal Antibody
Supplier: R&D Systems IC1051A
This item is not returnable.
View return policy
Description
IgG Monoclonal specifically detects IgG in Rabbit samples. It is validated for Control.Specifications
IgG | |
60024B | |
Control 10 uL/10^6 cells | |
Immunoglobulin G, ImmunoglobulinG | |
Protein A or G purified from cell culture supernatant | |
RUO | |
Selected for use as rabbit IgG control for flow cytometry applications. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG |
Monoclonal | |
APC | |
Supplied in a saline solution containing BSA and Sodium Azide. | |
Rabbit | |
200 Tests | |
3500 | |
Rabbit | |