Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG Rabbit, Clone: 60024B Isotype Control, Novus Biologicals™

Catalog No. MAB1050500
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1050500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB1050500 Supplier R&D Systems Supplier No. MAB1050500
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

IgG Monoclonal specifically detects IgG in Rabbit samples. It is validated for Control, CyTOF-ready.

Specifications

Antigen IgG
Applications CyTOF
Classification Monoclonal
Clone 60024B
Conjugate Unconjugated
Dilution Control, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene Alias Immunoglobulin G, ImmunoglobulinG
Host Species Rabbit
Immunogen KLH
Purification Method Protein A or G purified from cell culture supernatant
Quantity 500 μg
Regulatory Status RUO
Gene ID (Entrez) 3500
Test Specificity Selected for use as rabbit IgG isotype control for flow cytometry applications.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Rabbit
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.