Learn More
IgG1 Human, AP, Clone: , Isotype Control, Southern Biotech™
Shop All Southern Biotechnology Associates Products

Description
SouthernBiotech isotype controls and purified immunoglobulins are useful as negative controls and standards in a variety of immunoassays.
Specifications
Specifications
| Applications | ELISA |
| Concentration | Lot specific |
| Conjugate | Alkaline Phosphatase (AP) |
| Formulation | Supplied in a stock solution 50 mM Tris/1 mM MgCl2/50% glycerol, pH 8.0, containing NaN3 as preservative |
| Host Species | Human |
| Quantity | 1.0 mL |
| Regulatory Status | RUO |
| Primary or Secondary | Primary |
| Content And Storage | Store at 2-8°C |
| Isotype | IgG1 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.