Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG1, k Isotype Control, BU480, Clone: X40, BD Biosciences™

Catalog No. BDB565652 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB565652 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB565652 Supplier BD Biosciences Supplier No. 565652
Add to Cart
Add to Cart

Monoclonal Isotype Control

The X40 hybridoma was derived from the fusion of mouse Sp2/0-Ag 14 myeloma cells with spleen cells from BALB/c mice immunized with keyhole limpet hemocyanin (KLH). The hybridoma secretes monoclonal antibody that binds specifically to KLH, an antigen not expressed on human cells or human cell lines. It is validated for Flow Cytometry, Control
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG1, k
Applications Flow Cytometry
Classification Monoclonal
Clone X40
Conjugate Brilliant Ultraviolet 480
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Immunogen Keyhole limpet hemocyanin (KLH)
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Isotype Control (Primary)
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BV480 under optimum conditions, and unconjugated antibody and free BD Horizon BV480 were removed.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.