Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG1, Kappa Mouse, Alexa Fluor 488, Isotype Control, BD Pharmingen™

Catalog No. BDB567121 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB567121 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BDB567121 Supplier BD Biosciences Supplier No. 567121
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

IgG1, Kappa Monoclonal antibody specifically detects IgG1, Kappa. It is validated for Flow Cytometry, Isotype control
TRUSTED_SUSTAINABILITY

Specifications

Antigen igG1, Kappa
Applications Flow Cytometry
Classification Monoclonal
Clone X40
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 488
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias IgG1, kappa Control (Anti-KLH)
Host Species Mouse
Immunogen Keyhole limpet hemocyanin (KLH)
Quantity 0.1 mg
Regulatory Status RUO
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unreacted dye was removed.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.