Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG1, κ, Mouse, Alexa Fluor 647, Clone: MOPC-21, Isotype Control, BD Pharmingen™

Catalog No. BDB565571 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB565571 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB565571 Supplier BD Biosciences Supplier No. 565571
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Isotype Control

IgG1, κ Monoclonal antibody specifically detects IgG1, κ in samples. It is validated for Flow cytometry, Isotype control.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG1, κ
Applications Flow Cytometry
Classification Monoclonal
Clone MOPC-21 
Concentration 0.2mg/mL
Conjugate Alexa Fluor 647
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Form Purified
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Reseach Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.