Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG1, κ Mouse, Brilliant Ultraviolet 496, Clone: X40, Isotype Control, BD Horizon™

Catalog No. BDB612949 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB612949 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB612949 Supplier BD Biosciences Supplier No. 612949
Add to Cart
Add to Cart

Mouse Isotype Control

IgG1, κ Monoclonal isotype control is validated for Flow cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG1, κ
Applications Flow Cytometry
Classification Monoclonal
Clone X40
Concentration 0.2mg/mL
Conjugate Brilliant Ultraviolet 496
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Keyhole limpet hemocyanin (KLH)
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.