Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgG1, κ Mouse, PE-Cy™7, Clone: MOPC-21; Isotype Control, BD Pharmingen™
Mouse Monoclonal Isotype Control
Supplier: BD Biosciences 565573
Description
IgG1, κ Monoclonal antibody specifically detects IgG1, κ in samples. It is validated for Isotype control, Flow cytometry.
Specifications
IgG1, κ | |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
RUO | |
Primary |
Flow Cytometry | |
MOPC-21 | |
PE-Cyanine7 | |
IgG1, kappa; Ighg1, Igh-4, γg1; Igk-C, IGKC, Ig κ | |
50 μg | |
Cell Biology | |
Purified |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Reseach Use Only.
Spot an opportunity for improvement?Share a Content Correction