Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG1 Rat, PE/CY5.5, Clone: KLH/G1-2-2, Isotype Control, Southern Biotech™
SDP

Catalog No. OB011616 Shop All Southern Biotechnology Associates Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OB011616 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OB011616 Supplier Southern Biotechnology Associates Supplier No. 011616

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rat Monoclonal Isotype Control

SouthernBiotech isotype controls and purified immunoglobulins are useful as negative controls and standards in a variety of immunoassays.

Specifications

Applications Flow Cytometry
Classification Monoclonal
Clone KLH/G1-2-2
Concentration 0.1mg/mL
Conjugate PE-Cyanine5.5
Formulation Supplied in PBS/NaN3 and a stabilizing agent
Host Species Rat
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage Store at 2-8°C; Protect from light; Do not freeze
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.