Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgG2, κ Isotype Control, BUV615, BD Biosciences™
Monoclonal Isotype Control
Supplier: BD Biosciences 613012
Description
IgG2, κ Monoclonal antibody specifically detects IgG2, κ. It is validated for Flow Cytometry, Control
Specifications
IgG2, κ | |
Monoclonal | |
Brilliant Ultraviolet 615 | |
Anti-KLH | |
50 μg | |
Isotype Control (Primary) | |
IgG2 κ |
Flow Cytometry | |
B81-3 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Keyhole Limpet Hemocyanin | |
RUO | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction