Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG2, Lambda, Hamster, BV786, Clone: Ha4/8; Isotype Control, BD Horizon™

Catalog No. BDB565864 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB565864 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB565864 Supplier BD Biosciences Supplier No. 565864
Add to Cart
Add to Cart

Hamster Monoclonal Isotype Control

IgG2, Lambda Monoclonal antibody specifically detects IgG2, Lambda in samples. It is validated for Flow cytometry, Isotype control.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG2, Lambda
Applications Flow Cytometry
Classification Monoclonal
Clone Ha4/8 
Concentration 0.2mg/mL
Conjugate Brilliant Violet 786
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Gene Alias Anti-KLH
Host Species Hamster
Immunogen Keyhole limpet hemocyanin
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Form Purified
Isotype IgG2 γ1
Show More Show Less

For Reseach Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.