Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse IgG2a kappa Isotype Control (eBM2a), eBioscience™, Invitrogen™

Mouse Isotype Control

$70.50 - $130.00

Specifications

Antigen Mouse IgG2a kappa
Clone eBM2a
Concentration 0.5 mg/mL
Applications Control, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-130-48
Encompass_Preferred
View Documents
Invitrogen
14472481
50 μg
Each for $70.50
Only null left
Edge
Add to Cart
 
50-130-49
Encompass_Preferred
View Documents
Invitrogen
14472482
100 μg
Each for $130.00
Only null left
Edge
Add to Cart
 
Description

Description

Description: This is a monoclonal mouse IgG2a, kappa antibody. It is used as an isotype control for mouse IgG2a antibodies. Applications Reported: This mouse IgG2a isotype control has been reported for use in surface and intracellular flow cytometric analysis, immunocytochemistry, and immunohistochemistry. Applications Tested: This mouse IgG2a isotype control has been tested by flow cytometric analysis of human peripheral blood leukocytes and should be used at at the same concentration as the experimental antibody. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.

Mouse IgG2a is a subtype of immunoglobulin G antibody that is produced by B cells in response to an antigen. It is the second most abundant IgG subtype in the serum and plays a crucial role in the adaptive immune response. Mouse IgG2a is composed of two heavy chains and two light chains, and it is involved in opsonization, complement activation, and antibody-dependent cell-mediated cytotoxicity. It is also involved in the regulation of immune responses, including the activation of T cells and the inhibition of B cell activation. Mouse IgG2a is widely used in research and diagnostic applications, including ELISA, Western blotting, and immunohistochemistry. It is also used in therapeutic applications, such as immunotherapy and vaccine development.
Specifications

Specifications

Mouse IgG2a kappa
0.5 mg/mL
Unconjugated
Mouse
Not Applicable
IgG2; Immunoglobulin G; Immunoglobulin G2; ImmunoglobulinG; ImmunoglobulinG2
Isotype Control (Primary)
4° C
eBM2a
Control, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry
Liquid
RUO
PBS with 0.09% sodium azide; pH 7.2
IgG2a κ
Affinity chromatography
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.