Learn More
Rat IgG2a kappa Isotype Control (eBR2a), PE, eBioscience™, Invitrogen™
Rat Isotype Control
$100.00 - $287.65
Specifications
Antigen | Rat IgG2a kappa |
---|---|
Clone | eBR2a |
Conjugate | PE |
Form | Liquid |
Host Species | Rat |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-186-44
![]() |
Invitrogen
12432180 |
25 μg |
Each for $100.00
|
![]() |
|||||
50-112-9722
![]() |
Invitrogen
12432181 |
50 μg |
Each for $155.00
|
![]() |
|||||
50-112-9586
![]() |
Invitrogen
12432182 |
100 μg |
Each for $227.00
|
![]() |
|||||
50-112-9653
![]() |
Invitrogen
12432183 |
200 μg |
Each
for $287.65
Save 11% Reg : |
![]() |
|||||
50-112-9721
![]() |
Invitrogen
12432141 |
25 Tests |
Each for $112.00
|
![]() |
|||||
50-112-9612
![]() |
Invitrogen
12432142 |
100 Tests |
Each
for $189.65
Save 3% Reg : |
![]() |
|||||
Description
Description: The monoclonal rat IgG2a, kappa is useful as an isotype control immunoglobulin. Applications Reported: This rat IgG2a isotype control has been reported for use in flow cytometric analysis. Applications Tested: This Rat IgG2a K Isotype Control has been tested by flow cytometric analysis of mouse splenocytes and should be used at the same concentration as the experimental antibody. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.
Rat IgG2a is an immunoglobulin subclass that is commonly used in research and diagnostic applications. It is produced by the rat immune system and is known for its ability to activate complement and mediate antibody-dependent cell-mediated cytotoxicity (ADCC). Rat IgG2a has a molecular weight of approximately 160 kDa and is composed of two heavy chains and two light chains. It is often used as a control or reference antibody in experiments involving rat models, and is also commonly used in flow cytometry and immunohistochemistry applications.Specifications
Rat IgG2a kappa | |
PE | |
Rat | |
Not Applicable | |
IgG2a κ | |
Affinity chromatography |
eBR2a | |
Liquid | |
RUO | |
IgG2; Immunoglobulin G; Immunoglobulin G2; ImmunoglobulinG; ImmunoglobulinG2 | |
Isotype Control (Primary) | |
4° C, store in dark, DO NOT FREEZE! |
For Research Use Only.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.