Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGG2a, kappa Rabbit anti-Mouse, Clone: RM107, Abnova™

Catalog No. 89952799 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-952-799 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-952-799 Supplier Abnova Corporation Supplier No. MAB12767
Only null left
Add to Cart
Add to Cart

Rabbit monoclonal antibody raised against native mouse IgG2a (kappa).

Specifications

Antigen IGG2a, kappa
Applications ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone RM107
Conjugate Unconjugated
Description Rabbit monoclonal antibody raised against native mouse IgG2a (kappa).
Dilution ELISA (0.005 - 0.2 ug/mL) Flow Cytometry Immunocytochemistry (0.5 - 2 ug/mL) Immunohistochemistry (0.5 - 2 ug/mL) Immunoprecipitation Western Blot (0.1 - 0.5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (50% glycerol, 1% BSA, 0.09% sodium azide)
Gene Alias 1810060O09Rik/IGHG2A/Igh-1/MGC102662/MGC118125/MGC18977
Gene Symbols Igh-1a
Host Species Rabbit
Immunogen Native purified IgG from mouse serum.
Purification Method Protein A affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 380793
Test Specificity This antibody reacts to the Fab region of mouse IgG2a. No cross reactivity with mouse IgG2a, IgG1, IgG3, IgM, IgA, IgE, human IgG, rat IgG, or goat IgG.
Target Species Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.