Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgG2a Rat anti-Mouse, R-PE, Clone: SB84a, Southern Biotech™

Rat Monoclonal Secondary Antibody
$425.50 - $457.00
Specifications
Antigen | IgG2a |
---|---|
Clone | SB84a |
Concentration | 0.1mg/mL |
Applications | Flow Cytometry, ELISA, Immunoassay |
Classification | Monoclonal |
Description
SouthernBiotech monoclonal secondary antibodies offer high sensitivity and low background for use in a variety of applications.
Specifications
IgG2a | |
0.1mg/mL | |
Monoclonal | |
Rat | |
Mouse | |
Mouse IgG2a hybridoma | |
Secondary |
SB84a | |
Flow Cytometry, ELISA, Immunoassay | |
R-PE | |
RUO | |
Supplied in PBS/NaN3 and a stabilizing agent | |
IgG1 | |
Store at 2-8°C; Protect from light; Do not freeze |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title