Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG2b, κ Isotype Control Mouse RY610, Clone: 27-35, BD Horizon™

Catalog No. BDB571164 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB571164 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB571164 Supplier BD Biosciences Supplier No. 571164
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IgG2b, κ Isotype Control Monoclonal antibody specifically detects IgG2b, κ Isotype Control in samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG2b, κ Isotype Control
Applications Flow Cytometry
Classification Monoclonal
Clone 27-35
Concentration 0.2 mg/mL
Conjugate RY610
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias Anti-DNS; Anti-Dansyl; IgG2b kappa; Isotype Control (anti-Dansyl)
Host Species Mouse
Immunogen Dansyl (DNS) Hapten
Purification Method The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography.
Quantity 0.1 mg
Regulatory Status RUO
Target Species None
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.