Learn More
Mouse IgG2b kappa Isotype Control (eBMG2b), Functional Grade, eBioscience™, Invitrogen™
Mouse Isotype Control
$107.65 - $261.00
Specifications
Antigen | Mouse IgG2b kappa |
---|---|
Clone | eBMG2b |
Concentration | 1 mg/mL |
Applications | Control, Flow Cytometry, Functional Assay |
Conjugate | Functional Grade |
Description
Description: The monoclonal mouse IgG2b, kappa is useful as an isotype control immunoglobulin. Applications Reported: This mouse IgG2b isotype control has been reported for use in flow cytometric analysis. Applications Tested: This mouse IgG2b Isotype Control has been tested by flow cytometric analysis of normal human peripheral blood cells. Use isotype control at the same concentration as experimental antibody. Storage and handling: Use in a sterile environment. Filtration: 0.2 μm post-manufacturing filtered. Purity: Greater than 90%, as determined by SDS-PAGE. Endotoxin Level: Less than 0.001 ng/μg antibody, as determined by LAL assay. Aggregation: Less than 10%, as determined by HPLC.
Mouse IgG2b is a subtype of immunoglobulin G antibody that is produced by B cells in response to an antigen. It is the third most abundant IgG subtype in the serum and plays a crucial role in the adaptive immune response. Mouse IgG2b is composed of two heavy chains and two light chains, and it is involved in opsonization, complement activation, and antibody-dependent cell-mediated cytotoxicity. It is also involved in the regulation of immune responses, including the activation of T cells and the inhibition of B cell activation. Mouse IgG2b is widely used in research and diagnostic applications, including ELISA, Western blotting, and immunohistochemistry. It is also used in therapeutic applications, such as immunotherapy and vaccine development.Specifications
Mouse IgG2b kappa | |
1 mg/mL | |
Functional Grade | |
Mouse | |
Not Applicable | |
IgG2; Immunoglobulin G; Immunoglobulin G2; ImmunoglobulinG; ImmunoglobulinG2 | |
Isotype Control (Primary) | |
4° C |
eBMG2b | |
Control, Flow Cytometry, Functional Assay | |
Liquid | |
RUO | |
PBS with no preservative; pH 7.2 | |
IgG2b κ | |
Affinity chromatography |
For Research Use Only.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.