Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG2b, κ Mouse, RB545, Isotype Control, BD Horizon™

Catalog No. BDB569292 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB569292 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BDB569292 Supplier BD Biosciences Supplier No. 569292
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

This mouse IgG2b, κ isotype control is a monoclonal antibody, clone 27-35, that is specific for the dansyl (5-[dimethylamino] naphthalene-1-sulfonyl) hapten. The dansyl (DNS) hapten is not expressed on human cells or human cell lines. The 27-35 immunoglobulin was selected as an isotype control following testing that demonstrated low background staining on a variety of mouse and human tissues.
TRUSTED_SUSTAINABILITY

Specifications

Applications Flow Cytometry
Classification Monoclonal
Clone 27-35
Concentration 0.2 mg/mL
Conjugate RB545
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias Anti-DNS; Anti-Dansyl; IgG2b kappa; Isotype Control (anti-Dansyl)
Host Species Mouse
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unreacted dye was removed.
Form Purified
Isotype IgG2b κ
Excitation/Emission Excitation Source: Blue 488 nm, Excitation Max: 496 nm, Emission Max: 545 nm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.