Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgG2b Rat anti-Mouse, R-PE, Clone: SB74g, Southern Biotech™

Rat Monoclonal Secondary Antibody
$296.00 - $481.50
Specifications
Antigen | IgG2b |
---|---|
Clone | SB74g |
Concentration | 0.1mg/mL |
Applications | Flow Cytometry, ELISA |
Classification | Monoclonal |
Description
SouthernBiotech monoclonal secondary antibodies offer high sensitivity and low background for use in a variety of applications.
Specifications
IgG2b | |
0.1mg/mL | |
Monoclonal | |
Rat | |
Mouse | |
Mouse IgG2b hybridoma | |
Secondary |
SB74g | |
Flow Cytometry, ELISA | |
R-PE | |
RUO | |
Supplied in PBS/NaN3 and a stabilizing agent | |
IgG2b | |
Store at 2-8°C; Protect from light; Do not freeze |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title