Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG2c Antibody (RM223), Biotin, Novus Biologicals™
SDP

Catalog No. NBP262013B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP262013B 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP262013B Supplier Novus Biologicals Supplier No. NBP262013B
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

IgG2c Monoclonal specifically detects IgG2c in Mouse samples. It is validated for ELISA, Western Blot.

Specifications

Antigen IgG2c
Applications ELISA, Western Blot
Classification Monoclonal
Clone RM223
Concentration 1 mg/mL
Conjugate Biotin
Dilution Western Blot 0.5 ug/ml - 2 ug/ml, Flow Cytometry, ELISA 0.01 ug/ml - 0.2 ug/ml, Immunohistochemistry 0.5 - 2 ug/mL, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunocytochemistry 0.5 - 2 ug/mL
Formulation PBS, 50% Glycerol, 1% BSA with 0.09% Sodium Azide
Host Species Rabbit
Immunogen Mouse IgG2c
Purification Method Protein A purified
Quantity 50 μg
Primary or Secondary Secondary
Test Specificity This antibody reacts to the Fc region of mouse IgG2c.No cross reactivity with mouse IgG1, IgG2a, IgG2b,IgG3, IgM, IgA, IgE, human IgG, or rat IgG.
Target Species Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.