Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgG2c Goat anti-Mouse, Human ads, R-PE, Polyclonal, Southern Biotech™

Goat Polyclonal Secondary Antibody
$321.50 - $580.00
Specifications
Antigen | IgG2c |
---|---|
Concentration | 0.5mg/mL |
Applications | Flow Cytometry, ELISA |
Classification | Polyclonal |
Conjugate | R-PE |
Description
SouthernBiotech is a leading provider of polyclonal secondary antibodies offering high sensitivity and low background for use in a variety of applications.
Specifications
IgG2c | |
Flow Cytometry, ELISA | |
R-PE | |
RUO | |
Supplied in PBS/NaN3 and a stabilizing agent | |
IgG | |
Affinity Purified |
0.5mg/mL | |
Polyclonal | |
Goat | |
Mouse | |
Mouse IgG2c | |
Secondary | |
Store at 2-8°C; Protect from light; Do not freeze |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title