Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG3 Antibody (RM119), Biotin, Novus Biologicals™
SDP

Catalog No. NBP262125B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP262125B 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP262125B Supplier Novus Biologicals Supplier No. NBP262125B
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

IgG3 Monoclonal specifically detects IgG3 in Human samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Sandwich ELISA Capture, Sandwich ELISA Detection.

Specifications

Antigen IgG3
Applications ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Sandwich ELISA Capture, Sandwich ELISA Detection
Classification Monoclonal
Clone RM119
Concentration 1 mg/mL
Conjugate Biotin
Dilution Flow Cytometry, ELISA, Immunohistochemistry 0.5ug/ml - 2ug/ml, Immunocytochemistry/Immunofluorescence 0.5ug/ml - 2ug/ml, Immunocytochemistry 2 - 5 ug/mL
Formulation PBS, 50% Glycerol, 1% BSA with 0.09% Sodium Azide
Gene Alias DKFZp686H11213, FLJ39988, FLJ40036, FLJ40253, FLJ40587, FLJ40789, FLJ40834, IgG3, immunoglobulin heavy constant gamma 3 (G3m marker), MGC45809
Gene Symbols IGHG3
Host Species Rabbit
Immunogen Human IgG3
Purification Method Protein A purified
Quantity 50 μg
Primary or Secondary Secondary
Gene ID (Entrez) 3502
Test Specificity This antibody reacts to the heavy chain of human IgG3, This antibody does not cross react to any other IgG subclasses (IgG1, IgG2, or IgG4), and shows no cross reactivity IgM, IgA, IgD, IgE. This antibody does not react to monkey (Cyno or Rhesus) IgG, mouse IgG, rat IgG, or goat IgG.
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.