Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG4 Antibody (RM120), Biotin, R&D Systems™
SDP

Catalog No. NBP262126B Shop All Novus Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP262126B 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP262126B Supplier R&D Systems Supplier No. NBP262126B
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Rabbit Monoclonal Antibody

IgG4 Monoclonal specifically detects IgG4 in Human samples. It is validated for ELISA, Flow Cytometry, Immunocytochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Sandwich ELISA Capture, Sandwich ELISA Detection, Western Blot.

Specifications

Antigen IgG4
Applications ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Sandwich ELISA Capture, Sandwich ELISA Detection, Western Blot
Classification Monoclonal
Clone RM120
Concentration 1 mg/mL
Conjugate Biotin
Dilution Western Blot 0.5 ug/ml - 2 ug/ml, Flow Cytometry 1ug/ml - 4ug/ml, ELISA, Immunohistochemistry 1ug/mL - 10ug/mL, Immunocytochemistry/Immunofluorescence, Immunocytochemistry 1ug/mL - 10ug/mL
Formulation 50% Glycerol/PBS with 1% BSA with 0.09% Sodium Azide
Gene Alias Immunoglobulin G, Immunoglobulin G4, ImmunoglobulinG
Host Species Rabbit
Immunogen Peptide corresponding to the hinge region of Human IgG4
Purification Method Protein A purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Secondary
Test Specificity This antibody reacts to the heavy chain of human IgG4, This antibody does not cross react to any other IgG subclasses (IgG1, IgG2, or IgG3), and shows no cross reactivity to IgM, IgA, IgD, IgE. This antibody does not react to monkey (Cyno or Rhesus) IgG, mouse IgG, rat IgG, or goat IgG.
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.