Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IGHM Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598364
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598364 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598364 Supplier Invitrogen™ Supplier No. PA598364
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IGHM Polyclonal Antibody for Western Blot, IHC (P), ELISA

IgM is expressed intracellularly, during early stages of B lymphopoiesis, and then on the surface of more mature B cells in the bone marrow and peripheral B cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IGHM
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene IGHM
Gene Accession No. P01871
Gene Alias AGM1; IGHM; immunoglobulin heavy constant mu; MU; VH
Gene Symbols IGHM
Host Species Rabbit
Immunogen Recombinant Human Immunoglobulin heavy constant mu protein (45-178AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3507
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.