Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgM Antibody (II/41) - BSA Free, Novus Biologicals™

Rat Monoclonal Secondary Antibody
$206.50 - $499.50
Specifications
Antigen | Mouse IgM |
---|---|
Clone | II/41 |
Concentration | 0.5 mg/mL |
Dilution | Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Frozen |
Applications | Immunohistochemistry, Flow Cytometry, Immunohistochemistry (Frozen), ELISA |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB018529
![]() |
Novus Biologicals
NBP1433030.025MG |
0.025 mg |
Each for $206.50
|
|
|||||
NBP14330301
![]() |
Novus Biologicals
NBP1433030.1MG |
0.1 mg |
Each for $366.00
|
|
|||||
NBP1433035
![]() |
Novus Biologicals
NBP1433030.5MG |
0.5 mg |
Each for $499.50
|
|
|||||
Description
IgM Monoclonal specifically detects IgM in Mouse samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
Mouse IgM | |
0.5 mg/mL | |
Immunohistochemistry, Flow Cytometry, Immunohistochemistry (Frozen), ELISA | |
Unconjugated | |
RUO | |
Mouse | |
Constant region of heavy chain of IgM, DKFZp686I15196, DKFZp686I15212, FLJ00385, Hepatitis B virus receptor binding protein, Ig mu chain C region, IGHM, Immunoglobin heavy constant mu, Immunoglobulin heavy constant gamma 3, Immunoglobulin heavy mu chain, immunoglobulin mu, Immunoglobulin mu chain, Imunoglobulin heavy chain, MGC104996, MGC52291, mu, -region CH4 membrane bound domain, VH | |
IGHM | |
IgG2a κ | |
Affinity Purified |
II/41 | |
Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Frozen | |
Monoclonal | |
Rat | |
B Cell Development and Differentiation Markers, Immunology | |
Aqueous buffer, may contain carrier protein/stabilizer (proprietary). with 0.09% Sodium Azide | |
3507 | |
The immunogen for this antibody was IgM. | |
Secondary | |
Store at 4C. Do not freeze. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title