Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgM Antibody (II/41), PE/Cy7, Novus Biologicals™
SDP

Rat Monoclonal Secondary Antibody

$370.21 - $517.59

Specifications

Antigen Mouse IgM
Clone II/41
Concentration 0.2 mg/mL
Dilution Flow Cytometry
Applications Flow Cytometry
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP14294005
SDP
View Documents
Novus Biologicals
NBP1429400.05MG
0.05 mg
Each for $370.21
Only null left
Add to Cart
 
NBP14294001
SDP
View Documents
Novus Biologicals
NBP1429400.1MG
0.1 mg
Each for $517.59
Only null left
Add to Cart
 
Description

Description

IgM Monoclonal specifically detects IgM in Mouse samples. It is validated for Flow Cytometry.
Specifications

Specifications

Mouse IgM
0.2 mg/mL
Flow Cytometry
PE-Cyanine7
RUO
Mouse
Constant region of heavy chain of IgM, DKFZp686I15196, DKFZp686I15212, FLJ00385, Hepatitis B virus receptor binding protein, Ig mu chain C region, IGHM, Immunoglobin heavy constant mu, Immunoglobulin heavy constant gamma 3, Immunoglobulin heavy mu chain, immunoglobulin mu, Immunoglobulin mu chain, Imunoglobulin heavy chain, MGC104996, MGC52291, mu, -region CH4 membrane bound domain, VH
IGHM
IgG2a κ
Affinity Purified
II/41
Flow Cytometry
Monoclonal
Rat
Immunology
Aqueous buffer, may contain carrier protein/stabilizer (proprietary). with 0.09% Sodium Azide
3507
The immunogen for this antibody was IgM.
Secondary
Store at 4C. Do not freeze.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.