Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgM Antibody (II/41), PE, Novus Biologicals™
SDP

Catalog No. NBP14376305 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
0.1 mg
0.2 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP14376305 0.05 mg
NBP14376301 0.1 mg
NBP14376302 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NBP14376305 Supplier Novus Biologicals Supplier No. NBP1437630.05MG

Rat Monoclonal Secondary Antibody

IgM Monoclonal specifically detects IgM in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen Mouse IgM
Applications Flow Cytometry
Classification Monoclonal
Clone II/41
Concentration 0.2 mg/mL
Conjugate PE
Dilution Flow Cytometry
Formulation Aqueous buffer, may contain carrier protein/stabilizer (proprietary). with 0.09% Sodium Azide
Gene Alias Constant region of heavy chain of IgM, DKFZp686I15196, DKFZp686I15212, FLJ00385, Hepatitis B virus receptor binding protein, Ig mu chain C region, IGHM, Immunoglobin heavy constant mu, Immunoglobulin heavy constant gamma 3, Immunoglobulin heavy mu chain, immunoglobulin mu, Immunoglobulin mu chain, Imunoglobulin heavy chain, MGC104996, MGC52291, mu, -region CH4 membrane bound domain, VH
Gene Symbols IGHM
Host Species Rat
Immunogen The immunogen for this antibody was IgM.
Purification Method Affinity Purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline B Cell Development and Differentiation Markers, Immunology
Primary or Secondary Secondary
Gene ID (Entrez) 3507
Target Species Mouse
Content And Storage Store at 4C. Do not freeze.
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.