Learn More
IgM Goat anti-Mouse, Human ads, R-PE, Polyclonal, Southern Biotech™
Shop All Southern Biotechnology Associates Products

Description
SouthernBiotech is a leading provider of polyclonal secondary antibodies offering high sensitivity and low background for use in a variety of applications.
Specifications
Specifications
| Antigen | IgM |
| Applications | Flow Cytometry, ELISA |
| Classification | Polyclonal |
| Concentration | 0.5mg/mL |
| Conjugate | R-PE |
| Formulation | Supplied in PBS/NaN3 and a stabilizing agent |
| Host Species | Goat |
| Immunogen | Mouse IgM |
| Purification Method | Affinity Purified |
| Quantity | 0.5 mg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.