Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgM Heavy Chain Human Isotype Control, Novus Biologicals™

Human Isotype Control Antibody
Supplier: Novus Biologicals NBP196772
Description
IgM Heavy Chain Isotype Control antibody specifically detects IgM Heavy Chain in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
Native Human IgM, Fc5u | |
Unconjugated | |
0.1 M Tris Chloride, 0.5 M Sodium Chloride, pH 8.0 with 0.05% Sodium Azide | |
IGHM | |
Multi-step | |
RUO | |
Store at 4°C short term. For extended storage, add an equal volume of glycerol, aliquot and store at -20C or below. Avoid repeated freeze-thaw cycles. |
Western Blot, ELISA, Immunohistochemistry, PAGE | |
Western Blot 1:100-1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, SDS-Page | |
Constant region of heavy chain of IgM, Ig Mu Chain C Region BOT, Ig Mu Chain C Region GAL, Ig Mu Chain C Region OU, immunoglobulin heavy constant mu, Immunoglobulin mu chain | |
Human | |
1 mg | |
No reaction was observed against anti-Trypsin, anti-human IgG F(ab')2, anti-human IgG F(c), anti-human Kappa or anti-human Lambda. | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction