Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IgM Monoclonal Antibody (eB121-15F9), PE, eBioscience™, Invitrogen™
GREENER_CHOICE

Catalog No. 5010958
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-109-58 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-109-58 Supplier Invitrogen™ Supplier No. 12589082
Add to Cart
Edge
Add to Cart

Rat Monoclonal Antibody

Description: The eB121-15F9 monoclonal antibody reacts with the mu heavy chain of mouse IgM. It does not react with other classes of mouse immunoglobulin including IgD, IgG or IgA. IgM is expressed intracellularly, during early stages of B lymphopoiesis, and then on the surface of more mature B cells in the bone marrow and peripheral B cells. Fluorochrome conjugated eB121-15F9 can be used as a detection secondary for mouse IgM. Applications Reported: eB121-15F9 has been reported for use in flow cytometric analysis to detect the surface IgM on mouse B cells and as a second step to detect primary mouse IgMs. Applications Tested: The eB121-15F9 antibody has been tested by flow cytometric analysis to detect the surface IgM on mouse B cells and can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.

Mouse IgM is an immunoglobulin M antibody that is produced by B cells in response to an antigen. It is the first antibody produced during an immune response and is the largest immunoglobulin in the serum. Mouse IgM is composed of five heavy chains and five light chains, and it is involved in complement activation and agglutination of antigens. It plays a crucial role in the innate immune response and is important for neutralizing pathogens before the adaptive immune response is fully activated. Mouse IgM is widely used in research and diagnostic applications, including ELISA, Western blotting, and immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mouse IgM
Applications Flow Cytometry
Classification Monoclonal
Clone eB121-15F9
Concentration 0.2 mg/mL
Conjugate PE
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Ighm
Gene Accession No. P01872
Gene Alias AGM1; AI326478; Igh6; Igh-6; Ighm; Igh-M; Igm; immunoglobulin heavy constant mu; MU; muH; VH
Gene Symbols Ighm
Host Species Rat
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16019
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.