Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Ignenous Rock Set
SDPSCI_ED

Catalog No. S89552 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Grade:
5 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S89552 5 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy

Prop 65 Warning: Cancer and reproductive harm

Catalog No. S89552 Supplier United Scientific™ Supplier No. UNIGNRC01
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Prop 65 Warning: Cancer and reproductive harm

The United Scientific™ Ignenous Rock Set explores types of rocks that form through the cooling and solidification of magma or lava.

The United Scientific Ignenous Rock Set explores types of rocks that form through the cooling and solidification of magma or lava. Igneous rocks may form with or without crystallization, either below the surface as intrusive (plutonic) rocks or on the surface as extrusive (volcanic) rocks.

  • The set includes Andesite, Basalt, Diorite, Dolerite, Gabbro, Granite Black, Granite Porphyry, Granite Red, Granite White, Kimberlite, Obsidian Black, Obsidian Green, Pumice, And Rhyolite
  • Average sizes are 2 x 3 x 1.5 cm

Specifications

Grade 5 to 12
Format Kit
Product Type Rock Set
Includes 15 Sets
Languages English
Subject Geology
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.