Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IGSF3 Monoclonal Antibody (503621)
GREENER_CHOICE

Catalog No. PIMA524216 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524216 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524216 Supplier Invitrogen™ Supplier No. MA524216
  • $593.00
    $489.65 / Each of 1

    Save $103.35

    17% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

In direct ELISAs, no cross-reactivity with recombinant human (rh) IGSF4, 4B, 4C, 4D or 8 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.

IGSF3 (immunoglobulin superfamily, member 3), also known as V8 or EWI-3, is a 1,214 amino acid protein. Widely expressed with predominant expression in kidney, placenta and lung, IGSF3 localizes to the membrane and contains an N-terminal signal peptide, eight immunoglobulin (Ig) domains and a transmembrane segment. IGSF3 exhibits strong sequence and structural similarity to CD101 (32% identity), a leukocyte surface protein with seven Ig domains that is believed to play a role in T-cell activation. Despite the structural similarities between IGSF3 and CD101, IGSF3 is not expressed in peripheral blood lymphocytes and does not appear to participate in an immune function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IGSF3
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone 503621
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene IGSF3
Gene Accession No. O75054
Gene Alias 1700016K10Rik; 2810035F16Rik; 4833439O17Rik; EWI3; EWI-3; Glu-Trp-Ile EWI motif-containing protein 3; Igsf3; immunoglobin superfamily, member 3; immunoglobulin superfamily member 3; immunoglobulin superfamily, member 3; Kiaa0466; LCDD; mKIAA0466; RP4-781D12.1; V8
Gene Symbols IGSF3
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human IGSF3 Gln20-Ala1125.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3321
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.