Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iguratimod 98.0+%, TCI America™
SDP

Catalog No. I094525MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I094525MG 25 mg
I0945250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I094525MG Supplier TCI America Supplier No. I094525MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 123663-49-0
Molecular Formula C17H14N2O6S
Molecular Weight (g/mol) 374.367
MDL Number MFCD00882374
InChI Key ANMATWQYLIFGOK-UHFFFAOYSA-N
Synonym N-(3-Formamido-4-oxo-6-phenoxy-4H-chromen-7-yl)methanesulfonamide, N-[7-(Methanesulfonamido)-4-oxo-6-phenoxy-4H-chromen-3-yl]formamide
PubChem CID 124246
IUPAC Name N-[7-(methanesulfonamido)-4-oxo-6-phenoxychromen-3-yl]formamide
SMILES CS(=O)(=O)NC1=C(C=C2C(=C1)OC=C(C2=O)NC=O)OC3=CC=CC=C3

Specifications

Melting Point 240°C
Color White-Yellow
Quantity 25 mg
Formula Weight 374.37
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Iguratimod
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.