Learn More
Enzo Life Sciences IHC diluent (primary Ab) (250ml)
SDP
Supplier:  Enzo Life Sciences ADI9502440250
Encompass_Preferred
IHC diluent (primary Ab) can be used as a diluent to prepare primary antibodies and negative controls. It can be used with any primary antibody irrespective of its source of origin. Primary antibody diluent is specifically designed to stabilize primary antibodies and reduce nonspecific background staining observed during immunostaining. Formulation: Liquid. Primary antibody diluent. Long Term Storage: +4°C. Applications: IHC .
Catalog No. 50-200-7120
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.