Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IIP45 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501733000 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-3000 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-3000 Supplier Proteintech Group Inc Supplier No. 206301AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MIIP inhibits glioma cells invasion and down-regulates adhesion-and motility-associated genes such as NFKB2 and ICAM1. MIIP exhibits opposing effects to IGFBP2 on cell invasion.

Specifications

Antigen IIP45
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.45 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene MIIP
Gene Accession No. Q5JXC2
Gene Alias IGFBP2 binding protein, IIP45, invasion inhibitory protein 45, MIIP
Gene Symbols MIIP
Host Species Rabbit
Immunogen IIP45 Fusion Protein Ag14609
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60672
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.