Learn More
Selleck Chemical LLC iJMJD6-E1315-25MG
SDP
Supplier:  Selleck Chemical LLC E131525MG
Encompass_Preferred
iJMJD6(WL12) is an inhibitor of arginine demethylase JMJD6 with an IC50 value of 0 22 M
Catalog No. 50-313-7293