Learn More
Sigma Aldrich Fine Chemicals Biosciences AccurisTM Analytical Balan1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z7413461EA
Encompass
Accuris(TM) Analytical balances incorporate electromagnetic sensor technology and precision mechanical components to offer superior weight determinations to a fraction of a milligram. The glass draft shield enclosing the weighing chamber has sliding doors on the sides and top for easy access to the weigh pan.Leveling feet on the rear of the balance extend laterally to provide stability and a sensitive leveling bubble aids in the leveling process.
Catalog No. 50-290-8569
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.