Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IKAROS Monoclonal Antibody (2A9-mIkaros), eBioscience™, Invitrogen™
GREENER_CHOICE

Catalog No. p-7069460
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-245-566 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-245-566 Supplier Invitrogen™ Supplier No. 14578082
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Description: This 2A9-mIkaros monoclonal antibody reacts with mouse Ikaros, the 70-kDa founding member of the Ikaros family of C2H2-type zinc finger transcription factors. Ikaros is expressed in hematopoietic progenitor cells, thymocytes, mature T and B cells, monocytes, macrophages, and natural killer cells. This protein has also been shown to be expressed in non-immune cells such as neurons. Studies indicate that Ikaros can interact with other family members, namely Aiolos and Eos. Ikaros is involved in thymocyte development, B cell differentiation, cell cycle control, chromatin remodeling, and tumor suppression. Moreover, Ikaros has been implicated in leukemia and lymphoma. Ikaros has been demonstrated to regulate expression of Tdt, CD8a, and Notch target genes. At least eight Ikaros isoforms exist due to alternative splicing. The 2A9-mIkaros antibody has been reported to recognize Ikaros isoform VI (Ik-6), which is the longest and most predominant. Applications Reported: This 2A9-mIkaros antibody has been reported for use in western blotting. Applications Tested: This 2A9-mIkaros antibody has been tested by western blot of reduced mouse splenocytes. This can be used at less than or equal to 5 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.

DNA-binding protein IKAROS functions in the specification and the maturation of the T lymphocyte. It also interacts with a critical control element in the terminal deoxynucleotidyl transferase promoter as well as with the promoters for other genes expressed during early stages of B-cell and T-cell development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IKAROS
Applications Western Blot
Classification Monoclonal
Clone 2A9-mIkaros
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene IKZF1
Gene Accession No. Q03267
Gene Alias 5832432G11Rik; CLL-associated antigen KW-6; CVID13; DNA-binding protein Ikaros; hIk-1; hlk-1; Hs.54452; IK1; IKAROS; IKAROS family zinc finger 1; IKAROS family zinc finger 1 (Ikaros); ikaros family zinc finger protein 1; IKZF1; KAROS family zinc finger 1 (Ikaros); LYF1; LyF-1; Lymphoid transcription factor LyF-1; mKIAA4227; PPP1R92; PRO0758; protein phosphatase 1, regulatory subunit 92; RGD1562979; Zfpn1a1; zinc finger containing transcription factor; zinc finger protein, subfamily 1A, 1 (Ikaros); Znfn1a1
Gene Symbols IKZF1
Host Species Mouse
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22778
Target Species Mouse
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.