Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IL-1 alpha Monoclonal Antibody (ALF-161), PE, eBioscience™, Invitrogen™
GREENER_CHOICE

Catalog No. 5011118
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-111-18 100 μg
50-111-17 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-111-18 Supplier Invitrogen™ Supplier No. 12701182
  • $324.00
    $243.65 / Each of 1

    Save $80.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Armenian Hamster Monoclonal Antibody

Description: The ALF-161 antibody reacts with mouse interleukin-1alpha (IL-1alpha). Mouse IL-1alpha, also called Lymphocyte Activating Factor (LAF), Endogenous Pyrogen (EP), Leukocyte Endogenous Mediator (LEM), and Mononuclear Cell Factor (MCF), is a 17 kDa factor produced by a wide variety of cells, including macrophages, dendritic cells, T and B cells. IL-1alpha is mostly cell-associated, with 23% amino acid homology with IL-1beta. The immune regulatory role of IL-1alpha is exerted on a wide range of cells including lymphocytes, epithelial cells and fibroblasts. In vivo, it induces hypotension, fever, and acute phase response. Applications Reported: The ALF-161 antibody has been reported for use in ELISA capture, ELISPOT capture, neutralization, and intracellular staining for flow cytometry. Applications Tested: This ALF-161 antibody is offered in 2 formats: - μg size: Can can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. - test size: has been pre-titrated can be used at test size: 5 μL (0.25μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL.

IL-1 alpha (Interleukin-1 alpha) is a proinflammatory cytokine expressed by monocytes, macrophages, and dendritic cells. IL-1 alpha is coded by the IL1A gene and signals through two receptors, IL-1RI and IL-1RII, both of which are shared with IL-1 beta. IL1A is located on the q arm on chromosome 2 at position 13. IL-1 alpha plays an important role in innate host defense by triggering the production of other proinflammatory cytokines in target cells and initiating acute-phase responses. IL-1 alpha activity can be moderated by IL-1 Receptor Antagonist (IL-1RA), a protein produced by many cell types that blocks receptor binding through competitive inhibition. IL-1 alpha is a pleiotropic cytokine involved in various immune responses, inflammatory processes, and hematopoiesis. Further, IL-1 alpha is produced by monocytes and macrophages as a proprotein, which is proteolytically processed and released in response to cell injury and induces apoptosis. The IL1A gene and eight other interleukin 1 family genes form a cytokine gene cluster on chromosome 2. IL1 alpha regulates the activities of NF-kappaB and mitogen-activated protein kinases. IL-1 alpha can stimulate the expression of IL6 and cyclooxygenase-2 (PTGS2/COX-2), as well as enhance the production of nitric oxide (NO). High levels of IL-1 alpha are associated with several chronic inflammatory diseases including rheumatoid arthritis, psoriasis and multiple sclerosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-1 alpha
Applications Flow Cytometry
Classification Monoclonal
Clone ALF-161
Concentration 0.2 mg/mL
Conjugate PE
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Il1a
Gene Accession No. P01582
Gene Alias CTLA8; hematopoietin-1; IL 1 a; IL 1α; IL1; IL-1 alpha; IL17; IL1A; Il-1a; IL-1alpha; IL1-ALPHA; IL1F1; IL1α; ILN; Interleukin; interleukin 1 alpha; interleukin 1, alpha; interleukin 17; interleukin 1a; interleukin 1-alpha; interleukin 1-alpha (AA 1 - 152); Interleukin1 alpha; interleukin-1 alpha; interleukin-1 alpha precursor; Interleukin-17A; preinterleukin 1 alpha; pre-interleukin-1 alpha; pro-interleukin-1-alpha; RP23-160G19.8
Gene Symbols Il1a
Host Species Armenian Hamster
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16175
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.